missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetyl chloride, 98+%

Catalog No. AAA15051AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15051AE 100 mL
AAA15051AP 500 mL
AAA150510F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15051AE Supplier Thermo Scientific Chemicals Supplier No. A15051AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3282-30-2 | C5H9ClO | 120.58 g/mol

Trimethylacetyl chloride is used as a precursor in the preparation of tert-butyl peroxypivalate, guttiferon A derivatives, which is potential for the treatment of malaria. It is used as a raw material in the production of synthetic acidamide medicament and phenol ester medicament. In addition to this, it is used for the synthesis of active pharmaceutical ingredients such as aminobenzylpenicilin, cephalexin, cefazolin, dipivefrin and dipivalyl epinephrine. It is also used in heavy polymerization, N-acylating agent for amines, Schiff bases, pyrrolidinones as well as an O-acylating agent for alcohols, lactones and saccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetyl chloride is used as a precursor in the preparation of tert-butyl peroxypivalate, guttiferon A derivatives, which is potential for the treatment of malaria. It is used as a raw material in the production of synthetic acidamide medicament and phenol ester medicament. In addition to this, it is used for the synthesis of active pharmaceutical ingredients such as aminobenzylpenicilin, cephalexin, cefazolin, dipivefrin and dipivalyl epinephrine. It is also used in heavy polymerization, N-acylating agent for amines, Schiff bases, pyrrolidinones as well as an O-acylating agent for alcohols, lactones and saccharides.

Solubility
Miscible with acetonitrile.

Notes
Moisture sensitive. Incompatible withalcohols, oxidizing agents and strong bases.

Chemical Identifiers

CAS 3282-30-2
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000709
InChI Key JVSFQJZRHXAUGT-UHFFFAOYSA-N
Synonym pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride
PubChem CID 62493
IUPAC Name 2,2-dimethylpropanoyl chloride
SMILES CC(C)(C)C(Cl)=O

Specifications

Melting Point -57°C
Density 0.985
Boiling Point 105°C to 106°C
Flash Point 8°C (46°F)
Odor Pungent
Refractive Index 1.412
Quantity 100 mL
UN Number UN2438
Beilstein 385668
Sensitivity Moisture sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 120.58
Percent Purity ≥98%
Chemical Name or Material Trimethylacetyl chloride
Show More Show Less
Hazard Category H225-H290-H302-H314-H330
Hazard Statement GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapour.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TRIMETHYLACETYL CHLORIDE
EINECSNumber 221-921-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only