Learn More
Trimethylacetic anhydride, 99%
CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol
$655.37 - $655.37
Chemical Identifiers
| CAS | 1538-75-6 |
|---|---|
| Molecular Formula | C10H18O3 |
| Molecular Weight (g/mol) | 186.25 |
| MDL Number | MFCD00008842 |
| InChI Key | PGZVFRAEAAXREB-UHFFFAOYSA-N |
| Synonym | pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride |
| PubChem CID | 15234 |
| IUPAC Name | 2,2-dimethylpropanoyl 2,2-dimethylpropanoate |
| SMILES | CC(C)(C)C(=O)OC(=O)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC162442500
|
Thermo Scientific Chemicals
162442500 |
250 mL | Glass bottle |
Each for $655.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1538-75-6 | |
| 186.25 | |
| PGZVFRAEAAXREB-UHFFFAOYSA-N | |
| 15234 | |
| CC(C)(C)C(=O)OC(=O)C(C)(C)C |
| C10H18O3 | |
| MFCD00008842 | |
| pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride | |
| 2,2-dimethylpropanoyl 2,2-dimethylpropanoate |
Specifications
| 1538-75-6 | |
| 0.9100g/mL | |
| 57°C | |
| 98.5% min. (GC) | |
| C10H18O3 | |
| [(CH3)3CCO]2O | |
| 250 mL | |
| 0.91 | |
| Solubility in water: decomposes in water | |
| CC(C)(C)C(=O)OC(=O)C(C)(C)C | |
| 186.25 | |
| 186.25 | |
| Liquid |
| Colorless | |
| 193.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4080 to 1.4100 | |
| MFCD00008842 | |
| 02,32 | |
| pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride | |
| PGZVFRAEAAXREB-UHFFFAOYSA-N | |
| 2,2-dimethylpropanoyl 2,2-dimethylpropanoate | |
| 15234 | |
| 99% | |
| Trimethylacetic anhydride |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-263-1
RUO – Research Use Only