missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic acid, 99%

CAS: 75-98-9 | C5H10O2 | 102.133 g/mol

Supplier:  Thermo Scientific Chemicals A107760C

 View more versions of this product

Catalog No. AAA107760C


Only null left
Explore available promotions

Description

Description

Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

Solubility
Soluble in alcohol, ether. Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications

Specifications

Trimethylacetic acid
31°C to 35°C
163°C to 165°C
Pungent
MFCD00004194
UN3261
14,7511
Soluble in alcohol, ether; Slightly soluble in water
CC(C)(C)C(=O)O
102.133
CHEBI:45133
99%
75-98-9
0.91
63°C (145°F)
C5H10O2
10,000 g
969480
pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
IUGYQRQAERSCNH-UHFFFAOYSA-N
2,2-dimethylpropanoic acid
6417
102.13

Chemical Identifiers

75-98-9
102.133
IUGYQRQAERSCNH-UHFFFAOYSA-N
6417
2,2-dimethylpropanoic acid
C5H10O2
MFCD00004194
pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
CHEBI:45133
CC(C)(C)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c

H302-H311-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 200-922-5

RTECSNumber : TO7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only