missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Trimellitate 95.0+%, TCI America™

$38.51 - $284.92

Chemical Identifiers

CAS 10-1-2459
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00043629
InChI Key MJHNUUNSCNRGJE-UHFFFAOYSA-N
Synonym trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
PubChem CID 17159
IUPAC Name trimethyl benzene-1,2,4-tricarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T122925G
View Documents
TCI America
T122925G
25 g
Each for $38.51
Only null left
 
T1229500G
View Documents
TCI America
T1229500G
500 g
Each for $284.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10-1-2459
252.22
MJHNUUNSCNRGJE-UHFFFAOYSA-N
17159
COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC
C12H12O6
MFCD00043629
trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
trimethyl benzene-1,2,4-tricarboxylate

Specifications

10-1-2459
189°C
MFCD00043629
trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC
252.22
252.22
Liquid
Yellow
C12H12O6
25 g
MJHNUUNSCNRGJE-UHFFFAOYSA-N
trimethyl benzene-1,2,4-tricarboxylate
17159
≥95.0% (GC)
Trimethyl Trimellitate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1371

TSCA : Yes

SDS
Documents

Documents