missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Trimellitate 95.0+%, TCI America™

Catalog No. T1229500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1229500G 500 g
T122925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1229500G Supplier TCI America Supplier No. T1229500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10-1-2459
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00043629
InChI Key MJHNUUNSCNRGJE-UHFFFAOYSA-N
Synonym trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
PubChem CID 17159
IUPAC Name trimethyl benzene-1,2,4-tricarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC

Specifications

Color Yellow
Boiling Point 189°C
Quantity 500 g
Formula Weight 252.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Trimellitate
EINECSNumber (3)-1371
TSCA Yes