missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Phosphonoacetate 96.0+%, TCI America™

Catalog No. P126525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P126525G 25 g
P1265250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P126525G Supplier TCI America Supplier No. P126525G

Chemical Identifiers

CAS 5927-18-4
Molecular Formula C5H11O5P
Molecular Weight (g/mol) 182.112
MDL Number MFCD00008452
InChI Key SIGOIUCRXKUEIG-UHFFFAOYSA-N
Synonym trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate
PubChem CID 80029
IUPAC Name methyl 2-dimethoxyphosphorylacetate
SMILES COC(=O)CP(=O)(OC)OC

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 g
Formula Weight 182.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Phosphonoacetate