missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl phosphonoacetate, 98%
CAS: 5927-18-4 | C5H11O5P | 182.11 g/mol
$93.03 - $93.03
Chemical Identifiers
| CAS | 5927-18-4 |
|---|---|
| Molecular Formula | C5H11O5P |
| Molecular Weight (g/mol) | 182.11 |
| InChI Key | SIGOIUCRXKUEIG-UHFFFAOYSA-N |
| Synonym | trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate |
| PubChem CID | 80029 |
| IUPAC Name | methyl 2-dimethoxyphosphorylacetate |
| SMILES | COC(=O)CP(=O)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC140230250
|
Thermo Scientific Chemicals
140230250 |
25 g | Glass bottle |
Each for $93.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)Chemical Identifiers
| 5927-18-4 | |
| 182.11 | |
| trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate | |
| methyl 2-dimethoxyphosphorylacetate |
| C5H11O5P | |
| SIGOIUCRXKUEIG-UHFFFAOYSA-N | |
| 80029 | |
| COC(=O)CP(=O)(OC)OC |
Specifications
| 5927-18-4 | |
| 1.2600g/mL | |
| >112°C | |
| 97.5% min. (GC) | |
| C5H11O5P | |
| (CH3O)2P(O)CH2CO2CH3 | |
| 02,442; 07,394; 01,314; 02,169; 01,1236; 02,171; 13,15 | |
| trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate | |
| COC(=O)CP(=O)(OC)OC | |
| 182.11 | |
| 182.11 | |
| Liquid |
| Colorless to Yellow | |
| 265.0°C to 268.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4360 to 1.4380 | |
| 25 g | |
| 1.26 | |
| SIGOIUCRXKUEIG-UHFFFAOYSA-N | |
| methyl 2-dimethoxyphosphorylacetate | |
| 80029 | |
| 98% | |
| Trimethyl phosphonoacetate |
Safety and Handling
EINECSNumber : 227-663-
RUO – Research Use Only