missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl phosphonoacetate, 98%

CAS: 5927-18-4 | C5H11O5P | 182.112 g/mol

Supplier:  Thermo Scientific Chemicals A1330122

 View more versions of this product

Catalog No. AAA1330122


Only null left
Explore available promotions

Description

Description

Trimethyl phosphonoacetate is used in the preparation of sarin A by intramolecular Mannich-type reactions. It is involved in the Horner-Wadsworth-Emmons olefination with aldehydes and ketones to give acrylic esters. It is also involved in oxa-Michael reactions, prenylation of oxindoles and heterocyclization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl phosphonoacetate is used in the preparation of sarin A by intramolecular Mannich-type reactions. It is involved in the Horner-Wadsworth-Emmons olefination with aldehydes and ketones to give acrylic esters. It is also involved in oxa-Michael reactions, prenylation of oxindoles and heterocyclization reactions.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Trimethyl phosphonoacetate
1.263
70°C (158°F)
1.437
100 g
trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate
SIGOIUCRXKUEIG-UHFFFAOYSA-N
methyl 2-dimethoxyphosphorylacetate
80029
98%
5927-18-4
119°C to 120°C (1 mmHg)
C5H11O5P
MFCD00008452
1865177
Slightly miscible with water.
COC(=O)CP(=O)(OC)OC
182.112
182.11

Chemical Identifiers

5927-18-4
182.112
SIGOIUCRXKUEIG-UHFFFAOYSA-N
80029
COC(=O)CP(=O)(OC)OC
C5H11O5P
MFCD00008452
trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate
methyl 2-dimethoxyphosphorylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 227-663-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only