missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Phosphite Solution, NMR Reference Standard, 1% in acetone-d6 (99.9 atom % D), Ultra-Thin Wall, MilliporeSigma™ Supelco™

Catalog No. 111017064
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017064 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017064 Supplier Merck Emd Millipore Supplier No. 7362288IN
Only null left

NMR Tube Size: 5 mm 8 in., Ultra-Thin Wall

Specifications

Packaging NMR Tube Size: 5 mm 8 in., Ultra-Thin Wall
Concentration 1% in acetone-d 6 (99.9 atom% D)
MDL Number MFCD00008350
Quantity 0.7 mL
Grade NMR Reference Standard