missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(pentafluoroethyl)silane 98.0+%, TCI America™

Catalog No. p-7137527
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30111G 1 g
T30115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30111G Supplier TCI America Supplier No. T30111G
Only null left

Chemical Identifiers

CAS 124898-13-1
Molecular Formula C5H9F5Si
Molecular Weight (g/mol) 192.204
MDL Number MFCD04116436
InChI Key MTPVUVINMAGMJL-UHFFFAOYSA-N
Synonym pentafluoroethyl trimethylsilane, trimethyl pentafluoroethyl silane, pentafluoroethyltrimethylsilane, trimethylpentafluoroethylsilane, trimethyl perfluoroethyl silane, trimethyl 1,1,2,2,2-pentafluoroethyl silane, silane, trimethyl pentafluoroethyl, silane,trimethyl 1,1,2,2,2-pentafluoroethyl, acmc-20aple, trimethylsilylpentafluoroethane
PubChem CID 2760820
IUPAC Name trimethyl(1,1,2,2,2-pentafluoroethyl)silane
SMILES C[Si](C)(C)C(C(F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 70°C
Quantity 1 g
UN Number 1993
Formula Weight 192.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(pentafluoroethyl)silane