missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-p-tolylsilane, 95%

Catalog No. p-7023884
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1788406 5 g
AAA1788414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1788406 Supplier Thermo Scientific Chemicals Supplier No. A1788406

CAS: 3728-43-6 | C10H16Si | 164.323 g/mol

It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away.

Chemical Identifiers

CAS 3728-43-6
Molecular Formula C10H16Si
Molecular Weight (g/mol) 164.323
MDL Number MFCD00014850
InChI Key QGHURGPPCGMAMZ-UHFFFAOYSA-N
Synonym trimethyl-p-tolylsilane, p-tolyltrimethylsilane, trimethyl 4-methylphenyl silane, trimethyl-p-tolyl silane, trimethyl p-tolyl silane, 4-trimethylsilyl toluene, silane, p-tolyl-trimethyl-,, 4-methylphenyl trimethylsilane
PubChem CID 298149
IUPAC Name trimethyl-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C)(C)C

Specifications

Boiling Point 191°C to 192°C
Quantity 5 g
UN Number UN1993
Flash Point 46°C (115°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 164.33
Percent Purity 97%
Refractive Index 1.491
Density 0.865
Chemical Name or Material Trimethyl-p-tolylsilane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only