missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthovalerate 97.0+%, TCI America™

Catalog No. p-7136532
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T098125ML 25 mL
T0981250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T098125ML Supplier TCI America Supplier No. T098125ML
Only null left

Chemical Identifiers

CAS 13820-09-2
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00008481
InChI Key XUXVVQKJULMMKX-UHFFFAOYSA-N
Synonym trimethyl orthovalerate, pentane, 1,1,1-trimethoxy, unii-w0x9mom6vz, trimethylorthovalerate, orthovaleric acid, trimethyl ester, ortho-n-valeric acid trimethyl ester, w0x9mom6vz, trimethyl-2-valerate, trimethyl ortho-n-valerate, 1,1,1-trimethoxy-pentan
PubChem CID 83755
IUPAC Name 1,1,1-trimethoxypentane
SMILES CCCCC(OC)(OC)OC

Specifications

Color Colorless
Boiling Point 164°C
Quantity 25 mL
UN Number 3272
Formula Weight 162.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthovalerate