missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoformate 98.0+%, TCI America™

Catalog No. O006825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O006825ML 25 mL
O0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O006825ML Supplier TCI America Supplier No. O006825ML

Chemical Identifiers

CAS 149-73-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.121
MDL Number MFCD00008483
InChI Key PYOKUURKVVELLB-UHFFFAOYSA-N
Synonym trimethyl orthoformate, methyl orthoformate, methane, trimethoxy, orthoformic acid, trimethyl ester, orthoformic acid methyl ester, orthomravencan methylnaty, methoxymethylal, methylester kyseliny orthomravenci, unii-xam28819yj, orthoformic acid trimethyl ester
PubChem CID 9005
IUPAC Name trimethoxymethane
SMILES COC(OC)OC

Specifications

Melting Point -53°C
Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 3272
Formula Weight 106.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthoformate
EINECSNumber (2)-0682&(9)-0878
RTECSNumber RM6650000
TSCA Yes