missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoformate, 99%, Spectrum™ Chemical

Catalog No. p-7166698
Change view
Click to view available options
Quantity:
500 g
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614566 500 g Amber Glass Bottle
18614565 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614566 Supplier Spectrum Chemical Mfg Cor Supplier No. TR124500GM
Only null left

Chemical Identifiers

CAS 149-73-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.3765-1.3815
Quantity 500 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.