missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl Orthoacetate 98.0+%, TCI America™
$57.75 - $248.34
Chemical Identifiers
| CAS | 1445-45-0 |
|---|---|
| Molecular Formula | C5H12O3 |
| Molecular Weight (g/mol) | 120.148 |
| MDL Number | MFCD00008477 |
| InChI Key | HDPNBNXLBDFELL-UHFFFAOYSA-N |
| Synonym | trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate |
| PubChem CID | 15050 |
| IUPAC Name | 1,1,1-trimethoxyethane |
| SMILES | CC(OC)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O011525ML
|
TCI America
O011525ML |
25 mL |
Each for $57.75
|
|
|||||
|
O0115500ML
|
TCI America
O0115500ML |
500 mL |
Each for $248.34
|
|
|||||
Chemical Identifiers
| 1445-45-0 | |
| 120.148 | |
| HDPNBNXLBDFELL-UHFFFAOYSA-N | |
| 15050 | |
| CC(OC)(OC)OC |
| C5H12O3 | |
| MFCD00008477 | |
| trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate | |
| 1,1,1-trimethoxyethane |
Specifications
| 1445-45-0 | |
| 108°C | |
| MFCD00008477 | |
| 3272 | |
| HDPNBNXLBDFELL-UHFFFAOYSA-N | |
| 1,1,1-trimethoxyethane | |
| 15050 | |
| ≥98.0% (GC) | |
| Trimethyl Orthoacetate |
| Colorless | |
| C5H12O3 | |
| 25 mL | |
| trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate | |
| CC(OC)(OC)OC | |
| 120.148 | |
| 120.15 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2544
TSCA : Yes