missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthoacetate 98.0+%, TCI America™

Catalog No. O011525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O011525ML 25 mL
O0115500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O011525ML Supplier TCI America Supplier No. O011525ML

Chemical Identifiers

CAS 1445-45-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.148
MDL Number MFCD00008477
InChI Key HDPNBNXLBDFELL-UHFFFAOYSA-N
Synonym trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate
PubChem CID 15050
IUPAC Name 1,1,1-trimethoxyethane
SMILES CC(OC)(OC)OC

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 3272
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthoacetate
EINECSNumber (2)-2544
TSCA Yes