missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthoacetate, 98+%

Catalog No. AC218185000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218185000 500 mL Glass bottle
AC218181000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218185000 Supplier Thermo Scientific Chemicals Supplier No. 218185000

CAS: 1445-45-0 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1445-45-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00008477
InChI Key HDPNBNXLBDFELL-UHFFFAOYSA-N
Synonym trimethyl orthoacetate, ethane, 1,1,1-trimethoxy, orthoacetic acid, trimethyl ester, methyl orthoacetate, orthoacetic acid trimethyl ester, ch3c och3 3, unii-04h7a3fk37, tmoa, trimethoxyethane, trimethyl-acetate
PubChem CID 15050
IUPAC Name 1,1,1-trimethoxyethane
SMILES CC(OC)(OC)OC

Specifications

Color Colorless to Brown
Density 0.9400g/mL
Boiling Point 107.0°C to 109.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3890
Linear Formula CH3C(OCH3)3
Quantity 500 mL
Beilstein 02, II, 128
Fieser 17,377; 01,834; 02,318
Specific Gravity 0.94
Solubility Information Solubility in water: hydrolysis. Other solubilities: forming methanol and acetic acid
Formula Weight 120.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Trimethyl orthoacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-892-9
TSCA TSCA

RUO – Research Use Only