missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Citrate 98.0+%, TCI America™

Catalog No. p-7124378
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C03685G 5 g
C036825G 25 g
2 options

Catalog No. C03685G

Supplier: TCI America C03685G

Only null left

Chemical Identifiers

CAS 1587-20-8
Molecular Formula C9H14O7
Molecular Weight (g/mol) 234.204
MDL Number MFCD00025889
InChI Key HDDLVZWGOPWKFW-UHFFFAOYSA-N
Synonym Citric Acid Trimethyl Ester
PubChem CID 74112
IUPAC Name trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES COC(=O)CC(CC(=O)OC)(C(=O)OC)O

Specifications

Melting Point 79°C
Color White
Boiling Point 176°C
Quantity 5 g
Formula Weight 234.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl Citrate