missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Citrate 98.0+%, TCI America™

Catalog No. C03685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03685G 5 g
C036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03685G Supplier TCI America Supplier No. C03685G
Only null left

Chemical Identifiers

CAS 1587-20-8
Molecular Formula C9H14O7
Molecular Weight (g/mol) 234.204
MDL Number MFCD00025889
InChI Key HDDLVZWGOPWKFW-UHFFFAOYSA-N
Synonym Citric Acid Trimethyl Ester
PubChem CID 74112
IUPAC Name trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES COC(=O)CC(CC(=O)OC)(C(=O)OC)O

Specifications

Melting Point 79°C
Color White
Boiling Point 176°C
Quantity 5 g
Formula Weight 234.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl Citrate