missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 4-phosphonocrotonate, (E)+(Z), 90+%

CAS: 86120-40-3 | C7H13O5P | 208.15 g/mol

$155.88 - $605.49

Chemical Identifiers

CAS 86120-40-3
Molecular Formula C7H13O5P
Molecular Weight (g/mol) 208.15
MDL Number MFCD00051564
InChI Key PESRINKNQQZURC-SNAWJCMRSA-N
Synonym methyl 4-dimethoxyphosphoryl but-2-enoate, trimethyl 4-phosphonocrotonate, methyl 2e-4-dimethoxyphosphoryl but-2-enoate, methyl 4-dimethoxyphosphorylbut-2-enoate, trimethyl 4-phosphonocrotonate, e + z, methyl e-4-dimethoxyphosphorylbut-2-enoate, 4-dimethoxyphosphinyl crotonic acid methyl ester, e-4-dimethoxy-phosphoryl-but-2-enoic acid methyl ester
PubChem CID 5702556
IUPAC Name methyl (E)-4-dimethoxyphosphorylbut-2-enoate
SMILES COC(=O)\C=C\CP(=O)(OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1239909
View Documents
Thermo Scientific Chemicals
A1239909
10 g
Each for $155.88
Only null left
 
AAA1239918
View Documents
Thermo Scientific Chemicals
A1239918
50 g
Each for $605.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86120-40-3
208.15
PESRINKNQQZURC-SNAWJCMRSA-N
5702556
COC(=O)\C=C\CP(=O)(OC)OC
C7H13O5P
MFCD00051564
methyl 4-dimethoxyphosphoryl but-2-enoate, trimethyl 4-phosphonocrotonate, methyl 2e-4-dimethoxyphosphoryl but-2-enoate, methyl 4-dimethoxyphosphorylbut-2-enoate, trimethyl 4-phosphonocrotonate, e + z, methyl e-4-dimethoxyphosphorylbut-2-enoate, 4-dimethoxyphosphinyl crotonic acid methyl ester, e-4-dimethoxy-phosphoryl-but-2-enoic acid methyl ester
methyl (E)-4-dimethoxyphosphorylbut-2-enoate

Specifications

86120-40-3
C7H13O5P
10 g
methyl 4-dimethoxyphosphoryl but-2-enoate, trimethyl 4-phosphonocrotonate, methyl 2e-4-dimethoxyphosphoryl but-2-enoate, methyl 4-dimethoxyphosphorylbut-2-enoate, trimethyl 4-phosphonocrotonate, e + z, methyl e-4-dimethoxyphosphorylbut-2-enoate, 4-dimethoxyphosphinyl crotonic acid methyl ester, e-4-dimethoxy-phosphoryl-but-2-enoic acid methyl ester
COC(=O)\C=C\CP(=O)(OC)OC
208.15
208.15
Trimethyl 4-phosphonocrotonate, (E)+(Z)
123°C to 124°C (1 mmHg)
MFCD00051564
4987133
PESRINKNQQZURC-SNAWJCMRSA-N
methyl (E)-4-dimethoxyphosphorylbut-2-enoate
5702556
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only