missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 4-phosphonocrotonate, (E)+(Z), 90+%

Catalog No. AAA1239909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1239909 10 g
AAA1239918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1239909 Supplier Thermo Scientific Chemicals Supplier No. A1239909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86120-40-3 | C7H13O5P | 208.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86120-40-3
Molecular Formula C7H13O5P
Molecular Weight (g/mol) 208.15
MDL Number MFCD00051564
InChI Key PESRINKNQQZURC-SNAWJCMRSA-N
Synonym methyl 4-dimethoxyphosphoryl but-2-enoate, trimethyl 4-phosphonocrotonate, methyl 2e-4-dimethoxyphosphoryl but-2-enoate, methyl 4-dimethoxyphosphorylbut-2-enoate, trimethyl 4-phosphonocrotonate, e + z, methyl e-4-dimethoxyphosphorylbut-2-enoate, 4-dimethoxyphosphinyl crotonic acid methyl ester, e-4-dimethoxy-phosphoryl-but-2-enoic acid methyl ester
PubChem CID 5702556
IUPAC Name methyl (E)-4-dimethoxyphosphorylbut-2-enoate
SMILES COC(=O)\C=C\CP(=O)(OC)OC

Specifications

Boiling Point 123°C to 124°C (1 mmHg)
Quantity 10 g
Beilstein 4987133
Formula Weight 208.15
Percent Purity ≥90%
Chemical Name or Material Trimethyl 4-phosphonocrotonate, (E)+(Z)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only