missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(2-pyridyl)tin 98.0+%, TCI America™

$374.68 - $1200.44

Chemical Identifiers

CAS 13737-05-8
Molecular Formula C8H13NSn
Molecular Weight (g/mol) 241.91
MDL Number MFCD01631312
InChI Key HDWHGDDNMJOCCU-UHFFFAOYSA-N
Synonym 2-Trimethylstannylpyridine
PubChem CID 11021057
IUPAC Name 2-(trimethylstannyl)pyridine
SMILES C[Sn](C)(C)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T17421G
View Documents
TCI America
T17421G
1 g
Each for $374.68
Only null left
 
T17425G
View Documents
TCI America
T17425G
5 g
Each for $1,200.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13737-05-8
241.91
HDWHGDDNMJOCCU-UHFFFAOYSA-N
11021057
C[Sn](C)(C)C1=CC=CC=N1
C8H13NSn
MFCD01631312
2-Trimethylstannylpyridine
2-(trimethylstannyl)pyridine

Specifications

13737-05-8
C8H13NSn
1 g
2-Trimethylstannylpyridine
C[Sn](C)(C)C1=CC=CC=N1
241.91
241.91
Liquid
Yellow
MFCD01631312
2788
HDWHGDDNMJOCCU-UHFFFAOYSA-N
2-(trimethylstannyl)pyridine
11021057
≥98.0% (T)
Trimethyl(2-pyridyl)tin
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'