missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(2-pyridyl)tin 98.0+%, TCI America™

Catalog No. T17425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17425G 5 g
T17421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17425G Supplier TCI America Supplier No. T17425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13737-05-8
Molecular Formula C8H13NSn
Molecular Weight (g/mol) 241.91
MDL Number MFCD01631312
InChI Key HDWHGDDNMJOCCU-UHFFFAOYSA-N
Synonym 2-Trimethylstannylpyridine
PubChem CID 11021057
IUPAC Name 2-(trimethylstannyl)pyridine
SMILES C[Sn](C)(C)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 5 g
UN Number 2788
Formula Weight 241.91
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethyl(2-pyridyl)tin
TSCA No
Recommended Storage Refrigerator