missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl(2-pyridyl)tin 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 2788 |
| Formula Weight | 241.91 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Trimethyl(2-pyridyl)tin |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |