missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-1-octylammonium bromide, 97%

Catalog No. AAL0861006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0861006 5 g
AAL0861014 25 g
AAL0861022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0861006 Supplier Thermo Scientific Chemicals Supplier No. L0861006

CAS: 2083-68-3 | C11H26BrN | 252.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2083-68-3
Molecular Formula C11H26BrN
Molecular Weight (g/mol) 252.24
MDL Number MFCD00051873
InChI Key XCOHAFVJQZPUKF-UHFFFAOYSA-M
Synonym n-octyltrimethylammonium bromide, octalone, octyltrimethylammonium bromide, n,n,n-trimethyloctan-1-aminium bromide, 1-octanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethyl-1-octanaminium bromide, trimethyloctylammonium bromide 6ci,7ci, n-octyltrimethylammoniumbromide, trimethyloctylamine, bromide, trimethyl octyl azanium bromide
PubChem CID 74964
ChEBI CHEBI:346954
IUPAC Name trimethyl(octyl)azanium;bromide
SMILES CCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 220°C to 222°C
Quantity 5 g
Beilstein 3628448
Sensitivity Hygroscopic
Formula Weight 252.24
Percent Purity 97%
Chemical Name or Material Trimethyl-1-octylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber RG8275000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only