missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,3,5-Benzenetricarboxylate 98.0+%, TCI America™

Supplier:  TCI America T270725G

 View more versions of this product

Catalog No. T270725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethyl 1,3,5-Benzenetricarboxylate
146°C
230°C
MFCD00008434
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
252.222
252.22
Crystalline Powder
2672-58-4
White
C12H12O6
25 g
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl benzene-1,3,5-tricarboxylate
75881
≥98.0% (GC)

Chemical Identifiers

2672-58-4
252.222
RGCHNYAILFZUPL-UHFFFAOYSA-N
75881
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
C12H12O6
MFCD00008434
trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
trimethyl benzene-1,3,5-tricarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions