missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,3,5-Benzenetricarboxylate 98.0+%, TCI America™

Catalog No. T27075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27075G 5 g
T270725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27075G Supplier TCI America Supplier No. T27075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2672-58-4
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.222
MDL Number MFCD00008434
InChI Key RGCHNYAILFZUPL-UHFFFAOYSA-N
Synonym trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
PubChem CID 75881
IUPAC Name trimethyl benzene-1,3,5-tricarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 146°C
Color White
Boiling Point 230°C
Quantity 5 g
Formula Weight 252.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl 1,3,5-Benzenetricarboxylate