missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,3,5-benzenetricarboxylate, 99%

Catalog No. p-3737672
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152070250 25 g Glass bottle
AC152071000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152070250 Supplier Thermo Scientific Chemicals Supplier No. 152070250
Only null left

CAS: 2672-58-4 | C12H12O6 | 252.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2672-58-4
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00008434
InChI Key RGCHNYAILFZUPL-UHFFFAOYSA-N
Synonym trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766
PubChem CID 75881
IUPAC Name trimethyl benzene-1,3,5-tricarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 144°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH3)3
Quantity 25 g
Beilstein 09, 979
Formula Weight 252.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate
Show More Show Less
EINECSNumber 220-215-5

RUO – Research Use Only