missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,2,3-Propanetricarboxylate 97.0+%, TCI America™

Catalog No. T24291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24291G 1 g
T24295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24291G Supplier TCI America Supplier No. T24291G

Chemical Identifiers

CAS 6138-26-7
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
MDL Number MFCD00040552
InChI Key IXINWTSBOAMACD-UHFFFAOYSA-N
Synonym 1,2,3-Propanetricarboxylic Acid Trimethyl Ester, Dimethyl 3-(Methoxycarbonyl)pentanedioate, 3-(Methoxycarbonyl)pentanedioic Acid Dimethyl Ester
PubChem CID 581486
IUPAC Name 1,2,3-trimethyl propane-1,2,3-tricarboxylate
SMILES COC(=O)CC(CC(=O)OC)C(=O)OC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 218.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl 1,2,3-Propanetricarboxylate