missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxyphenylsilane 98.0+%, TCI America™

Catalog No. P0857500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0857500ML 500 mL
P085725ML 25 mL
P0857100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0857500ML Supplier TCI America Supplier No. P0857500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2996-92-1
Molecular Formula C9H14O3Si
Molecular Weight (g/mol) 198.29
MDL Number MFCD00025689
InChI Key ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Synonym phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
PubChem CID 18137
IUPAC Name trimethoxy(phenyl)silane
SMILES CO[Si](OC)(OC)C1=CC=CC=C1

Specifications

Melting Point -25°C
Color Colorless
Boiling Point 131°C
Quantity 500 mL
Formula Weight 198.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxyphenylsilane
EINECSNumber (3)-2635
RTECSNumber VV5252000
TSCA Yes