missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxyphenylsilane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -25°C |
| Color | Colorless |
| Boiling Point | 131°C |
| Quantity | 100 mL |
| Formula Weight | 198.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Trimethoxyphenylsilane |
Safety and Handling
| EINECSNumber | (3)-2635 |
| RTECSNumber | VV5252000 |
| TSCA | Yes |