missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(propyl)silane 98.0+%, TCI America™

Catalog No. T180125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T180125ML 25 mL
T1801500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T180125ML Supplier TCI America Supplier No. T180125ML

Chemical Identifiers

CAS 1067-25-0
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00043026
InChI Key HQYALQRYBUJWDH-UHFFFAOYSA-N
Synonym trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane
PubChem CID 61254
IUPAC Name trimethoxy(propyl)silane
SMILES CCC[Si](OC)(OC)OC

Specifications

Color Colorless
Quantity 25 mL
UN Number 1993
Formula Weight 164.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(propyl)silane
EINECSNumber (2)-2052&(2)-2053
TSCA Yes