missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(p-tolyl)silane 97.0+%, TCI America™

Catalog No. T25605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25605G Supplier TCI America Supplier No. T25605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17873-01-7
Molecular Formula C10H16O3Si
Molecular Weight (g/mol) 212.32
MDL Number MFCD00048020
InChI Key XQEGZYAXBCFSBS-UHFFFAOYSA-N
PubChem CID 2760672
IUPAC Name trimethoxy-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](OC)(OC)OC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 5 g
Formula Weight 212.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(p-tolyl)silane
TSCA No
Recommended Storage Refrigerator