missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(methyl)silane 98.0+%, TCI America™

Catalog No. M066025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M066025ML 25 mL
M0660100ML 100 mL
M0660500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M066025ML Supplier TCI America Supplier No. M066025ML

Chemical Identifiers

CAS 1185-55-3
Molecular Formula C4H12O3Si
Molecular Weight (g/mol) 136.222
MDL Number MFCD00008342
InChI Key BFXIKLCIZHOAAZ-UHFFFAOYSA-N
Synonym methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
PubChem CID 14456
IUPAC Name trimethoxy(methyl)silane
SMILES CO[Si](C)(OC)OC

Specifications

Color Colorless
Boiling Point 103°C
Quantity 25 mL
UN Number 1993
Formula Weight 136.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(methyl)silane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV4650000
TSCA Yes