missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane 97.0+%, TCI America™
$149.13 - $538.71
Chemical Identifiers
| CAS | 85857-16-5 |
|---|---|
| Molecular Formula | C11H13F13O3Si |
| Molecular Weight (g/mol) | 468.286 |
| MDL Number | MFCD00153261 |
| InChI Key | BVQYIDJXNYHKRK-UHFFFAOYSA-N |
| Synonym | Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane |
| PubChem CID | 2776334 |
| IUPAC Name | trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
| SMILES | CO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OC)OC |
Chemical Identifiers
| 85857-16-5 | |
| 468.286 | |
| BVQYIDJXNYHKRK-UHFFFAOYSA-N | |
| 2776334 | |
| CO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OC)OC |
| C11H13F13O3Si | |
| MFCD00153261 | |
| Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane | |
| trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
Specifications
| 85857-16-5 | |
| 71°C | |
| MFCD00153261 | |
| Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane | |
| CO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OC)OC | |
| 468.286 | |
| 468.29 | |
| Liquid |
| Colorless | |
| C11H13F13O3Si | |
| 5 g | |
| BVQYIDJXNYHKRK-UHFFFAOYSA-N | |
| trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane | |
| 2776334 | |
| ≥97.0% (GC) | |
| Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane |