missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane 97.0+%, TCI America™

Catalog No. T356025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T356025G 25 g
T35605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T356025G Supplier TCI America Supplier No. T356025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85857-16-5
Molecular Formula C11H13F13O3Si
Molecular Weight (g/mol) 468.286
MDL Number MFCD00153261
InChI Key BVQYIDJXNYHKRK-UHFFFAOYSA-N
Synonym Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane
PubChem CID 2776334
IUPAC Name trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES CO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OC)OC

Specifications

Color Colorless
Boiling Point 71°C
Quantity 25 g
Formula Weight 468.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane