missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic Anhydride 98.0+%, TCI America™

Catalog No. p-7124706
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C095925G 25 g
C0959500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C095925G Supplier TCI America Supplier No. C095925G
Only null left

Chemical Identifiers

CAS 552-30-7
Molecular Formula C9H4O5
Molecular Weight (g/mol) 192.126
MDL Number MFCD00005925
InChI Key SRPWOOOHEPICQU-UHFFFAOYSA-N
Synonym trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
PubChem CID 11089
ChEBI CHEBI:53050
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O

Specifications

Melting Point 168°C
Color White
Boiling Point 245°C
Quantity 25 g
Formula Weight 192.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimellitic Anhydride
EINECSNumber (3)-1362
RTECSNumber DC2050000
TSCA Yes