missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic Acid 98.0+%, TCI America™

Catalog No. B004225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B004225G 25 g
B0042100G 100 g
B0042500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B004225G Supplier TCI America Supplier No. B004225G

Chemical Identifiers

CAS 528-44-9
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002470
InChI Key ARCGXLSVLAOJQL-UHFFFAOYSA-N
Synonym 1,2,4-benzenetricarboxylic acid, trimellitic acid, 1,2,4-tricarboxybenzene, 4-carboxyphthalic acid, 1,3,4-benzenetricarboxylic acid, 1,4,5-benzenetricarboxylic acid, 1,2,4-benzenetricarboxylicacid, unii-7nvy29mq5f, 7nvy29mq5f, dsstox_cid_1487
PubChem CID 10708
ChEBI CHEBI:166055
IUPAC Name benzene-1,2,4-tricarboxylic acid
SMILES OC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

Melting Point 231°C
Color White
Quantity 25 g
Formula Weight 210.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimellitic Acid
EINECSNumber (3)-1279
RTECSNumber DC1980000
TSCA Yes