missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimagnesium Dicitrate Nonahydrate 98.0+%, TCI America™
$46.60 - $160.30
Chemical Identifiers
| CAS | 153531-96-5 |
|---|---|
| Molecular Formula | C12H34Mg3O23 |
| Molecular Weight (g/mol) | 619.296 |
| MDL Number | MFCD00036422 |
| InChI Key | YMNRVQRRBHEJPL-UHFFFAOYSA-N |
| PubChem CID | 131848878 |
| IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;magnesium;nonahydrate |
| SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.[Mg].[Mg].[Mg] |
Chemical Identifiers
| 153531-96-5 | |
| 619.296 | |
| YMNRVQRRBHEJPL-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid;magnesium;nonahydrate |
| C12H34Mg3O23 | |
| MFCD00036422 | |
| 131848878 | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.[Mg].[Mg].[Mg] |
Specifications
| 153531-96-5 | |
| C12H34Mg3O23 | |
| 25 g | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.[Mg].[Mg].[Mg] | |
| 619.296 | |
| 451.11 | |
| Crystalline Powder |
| White | |
| MFCD00036422 | |
| YMNRVQRRBHEJPL-UHFFFAOYSA-N | |
| 2-hydroxypropane-1,2,3-tricarboxylic acid;magnesium;nonahydrate | |
| 131848878 | |
| ≥98.0% (T) | |
| Trimagnesium Dicitrate Nonahydrate |
Safety and Handling
EINECSNumber : (2)-1323
RTECSNumber : TZ8552900
TSCA : No