missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trilostane

Numéro de catalogue. AC464382500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC464382500 250 mg
AC464380010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC464382500 Fournisseur Thermo Scientific Chemicals Code fournisseur 464382500
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 13647-35-3
Molecular Formula C20H27NO3
Molecular Weight (g/mol) 329.44
InChI Key KVJXBPDAXMEYOA-CXANFOAXSA-N
IUPAC Name (1S,2R,6R,8S,11S,12S,15S,16S)-5,15-dihydroxy-2,16-dimethyl-7-oxapentacyclo[9.7.0.0²,⁸.0⁶,⁸.0¹²,¹⁶]octadec-4-ene-4-carbonitrile
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@@]45O[C@@H]4C(O)=C(C[C@]35C)C#N)[C@@H]1CC[C@@H]2O

Spécifications

Melting Point 284.0°C
Quantity 250 mg
Formula Weight 329.43
Chemical Name or Material Trilostane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 237-133-0

RUO – Research Use Only