missing translation for 'onlineSavingsMsg'
Learn More

Trilaurin 98.0+%, TCI America™

Supplier:  TCI America G0087500G

 View more versions of this product

Catalog No. G0087500G


Only null left

Description

Description

Specifications

Specifications

Trilaurin
47°C
C39H74O6
500 g
VMPHSYLJUKZBJJ-UHFFFAOYSA-N
2,3-di(dodecanoyloxy)propyl dodecanoate
10851
639.02
Crystalline Powder
538-24-9
White-Yellow
MFCD00026559
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
639.015
CHEBI:77389
≥98.0% (GC)

Chemical Identifiers

538-24-9
639.015
VMPHSYLJUKZBJJ-UHFFFAOYSA-N
10851
2,3-di(dodecanoyloxy)propyl dodecanoate
C39H74O6
MFCD00026559
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
CHEBI:77389
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0669

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions