missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trilaurin 98.0+%, TCI America™
Supplier: TCI America G0087100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| C39H74O6 | |
| MFCD00026559 | |
| trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri | |
| CHEBI:77389 | |
| CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC |
Specifications
| Trilaurin | |
| 47°C | |
| C39H74O6 | |
| MFCD00026559 | |
| VMPHSYLJUKZBJJ-UHFFFAOYSA-N | |
| 2,3-di(dodecanoyloxy)propyl dodecanoate | |
| 10851 | |
| 639.02 | |
| Crystalline Powder |
| 538-24-9 | |
| White-Yellow | |
| 100 g | |
| trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri | |
| CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC | |
| 639.015 | |
| CHEBI:77389 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (2)-0669
TSCA : Yes