Learn More
Triisopropyl phosphite, 96%, Thermo Scientific Chemicals
$104.93 - $104.93
Chemical Identifiers
| CAS | 116-17-6 |
|---|---|
| Molecular Formula | C9H21O3P |
| Molecular Weight (g/mol) | 208.24 |
| InChI Key | SJHCUXCOGGKFAI-UHFFFAOYSA-N |
| Synonym | triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite |
| PubChem CID | 8304 |
| IUPAC Name | tripropan-2-yl phosphite |
| SMILES | CC(C)OP(OC(C)C)OC(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC372381000
|
Thermo Scientific Chemicals
372381000 |
100 g | Glass bottle |
Each for $104.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 116-17-6 | |
| 208.24 | |
| triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite | |
| tripropan-2-yl phosphite |
| C9H21O3P | |
| SJHCUXCOGGKFAI-UHFFFAOYSA-N | |
| 8304 | |
| CC(C)OP(OC(C)C)OC(C)C |
Specifications
| 116-17-6 | |
| 94°C to 95°C (66.6 hPa) | |
| Authentic | |
| Glass bottle | |
| 1.4095 to 1.4115 | |
| 100 g | |
| 01,1229; 16,36 | |
| triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite | |
| SJHCUXCOGGKFAI-UHFFFAOYSA-N | |
| tripropan-2-yl phosphite | |
| 8304 | |
| 96% | |
| Triisopropyl phosphite |
| 0.9140g/mL | |
| 62°C | |
| 95% min. (GC) | |
| C9H21O3P | |
| [(CH3)2CHO]3P | |
| 01,363 | |
| 0.914 | |
| Solubility in water: insoluble. | |
| CC(C)OP(OC(C)C)OC(C)C | |
| 208.24 | |
| 208.24 | |
| Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 204-130-
RUO – Research Use Only