missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl phosphite, 96%, Thermo Scientific™

Catalog No. AC372385000
Change view
Click to view available options
500g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC372385000 500g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC372385000 Supplier Thermo Scientific Chemicals Supplier No. 372385000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 116-17-6
Density 0.9140g/mL
Boiling Point 94°C to 95°C (66.6 hPa)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C9H21O3P
Linear Formula [(CH3)2CHO]3P
Refractive Index 1.4095 to 1.4115
Quantity 500g
Beilstein 01,363
Fieser 01,1229; 16,36
Specific Gravity 0.914
Synonym triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
Solubility Information Solubility in water: insoluble.
InChI Key SJHCUXCOGGKFAI-UHFFFAOYSA-N
SMILES CC(C)OP(OC(C)C)OC(C)C
IUPAC Name tripropan-2-yl phosphite
Molecular Weight (g/mol) 208.24
PubChem CID 8304
Formula Weight 208.24
Percent Purity 96%
Chemical Name or Material Triisopropyl phosphite
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
EINECSNumber 204-130-

RUO – Research Use Only