missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropoxyvanadium(V) Oxide 97.0+%, TCI America™
Supplier: TCI America T254825G
Specifications
| Triisopropoxyvanadium(V) Oxide | |
| -2°C | |
| 102°C | |
| MFCD00015017 | |
| vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n | |
| O=[V].CC(C)O.CC(C)O.CC(C)O | |
| 247.23 | |
| 244.20 | |
| Liquid |
| 5588-84-1 | |
| Yellow | |
| C9H24O4V | |
| 25 g | |
| JOUSPCDMLWUHSO-UHFFFAOYSA-N | |
| oxovanadium;propan-2-ol | |
| 79702 | |
| ≥97.0% (T) |
Chemical Identifiers
| 5588-84-1 | |
| 247.23 | |
| JOUSPCDMLWUHSO-UHFFFAOYSA-N | |
| 79702 | |
| O=[V].CC(C)O.CC(C)O.CC(C)O |
| C9H24O4V | |
| MFCD00015017 | |
| vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n | |
| oxovanadium;propan-2-ol |
Safety and Handling
EINECSNumber : (2)-3398
TSCA : Yes