missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine (mixture of isomer) 98.0+%, TCI America™

Catalog No. N010425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010425G 25 g
N0104500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N010425G Supplier TCI America Supplier No. N010425G

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.271
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O

Specifications

Color White
Boiling Point 305°C
Quantity 25 g
Formula Weight 191.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Triisopropanolamine (mixture of isomer)
EINECSNumber (2)-0310
RTECSNumber UB8750000
TSCA Yes