Learn More
Triisopropanolamine, 95%
CAS: 122-20-3 | C9H21NO3 | 191.271 g/mol
Supplier: Thermo Scientific Chemicals A199660C
Description
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMajor applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| Triisopropanolamine | |
| 48°C to 52°C | |
| >110°C (230°F) | |
| C9H21NO3 | |
| MFCD00004533 | |
| 1071570 | |
| triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891 | |
| SLINHMUFWFWBMU-UHFFFAOYSA-N | |
| 1-[bis(2-hydroxypropyl)amino]propan-2-ol | |
| 24730 | |
| 95% |
| 122-20-3 | |
| 189°C to 191°C (23 mmHg) | |
| Amine-like | |
| (CH3CHOHCH2)3N | |
| 10,000 g | |
| Hygroscopic | |
| Insoluble in water. Soluble ethanol,diethyl ether,chloroform (slightly ),and methanol (>500g/100g). | |
| CC(CN(CC(C)O)CC(C)O)O | |
| 191.271 | |
| 191.27 |
Chemical Identifiers
| 122-20-3 | |
| 191.271 | |
| SLINHMUFWFWBMU-UHFFFAOYSA-N | |
| 24730 | |
| CC(CN(CC(C)O)CC(C)O)O |
| C9H21NO3 | |
| MFCD00004533 | |
| triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891 | |
| 1-[bis(2-hydroxypropyl)amino]propan-2-ol |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 204-528-4
RTECSNumber : UB8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only