missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisobutylene (mixture of branched chain isomer) 90.0+%, TCI America™
$89.48 - $369.08
Chemical Identifiers
| CAS | 7756-94-7 |
|---|---|
| Molecular Formula | C12H24 |
| Molecular Weight (g/mol) | 168.324 |
| MDL Number | MFCD00060236 |
| InChI Key | DRHABPMHZRIRAH-UHFFFAOYSA-N |
| Synonym | Isobutene Trimer, Isobutylene Trimer |
| PubChem CID | 170244 |
| IUPAC Name | 2,4,4,6,6-pentamethylhept-2-ene |
| SMILES | CC(=CC(C)(C)CC(C)(C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T045625ML
|
TCI America
T045625ML |
25 mL |
Each for $89.48
|
|
|||||
|
T0456500ML
|
TCI America
T0456500ML |
500 mL |
Each for $369.08
|
|
|||||
Chemical Identifiers
| 7756-94-7 | |
| 168.324 | |
| DRHABPMHZRIRAH-UHFFFAOYSA-N | |
| 170244 | |
| CC(=CC(C)(C)CC(C)(C)C)C |
| C12H24 | |
| MFCD00060236 | |
| Isobutene Trimer, Isobutylene Trimer | |
| 2,4,4,6,6-pentamethylhept-2-ene |
Specifications
| 7756-94-7 | |
| 177°C | |
| MFCD00060236 | |
| 2324 | |
| DRHABPMHZRIRAH-UHFFFAOYSA-N | |
| 2,4,4,6,6-pentamethylhept-2-ene | |
| 170244 | |
| ≥90.0% (GC) | |
| Triisobutylene (mixture of branched chain isomer) |
| Colorless | |
| C12H24 | |
| 25 mL | |
| Isobutene Trimer, Isobutylene Trimer | |
| CC(=CC(C)(C)CC(C)(C)C)C | |
| 168.324 | |
| 168.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0032&(2)-0027
TSCA : Yes