missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisobutylene (mixture of branched chain isomer) 90.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 177°C |
| Quantity | 25 mL |
| UN Number | 2324 |
| Formula Weight | 168.32 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Triisobutylene (mixture of branched chain isomer) |
Safety and Handling
| EINECSNumber | (2)-0032&(2)-0027 |
| TSCA | Yes |