missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisobutylamine 98.0+%, TCI America™
$85.75 - $682.24
Chemical Identifiers
| CAS | 1116-40-1 |
|---|---|
| Molecular Formula | C12H27N |
| Molecular Weight (g/mol) | 185.36 |
| MDL Number | MFCD00040501 |
| InChI Key | IIFFFBSAXDNJHX-UHFFFAOYSA-N |
| PubChem CID | 14222 |
| IUPAC Name | tris(2-methylpropyl)amine |
| SMILES | CC(C)CN(CC(C)C)CC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T096625ML
|
TCI America
T096625ML |
25 mL |
Each for $85.75
|
|
|||||
|
T0966100ML
|
TCI America
T0966100ML |
100 mL |
Each for $212.51
|
|
|||||
|
T0966500ML
|
TCI America
T0966500ML |
500 mL |
Each for $682.24
|
|
|||||
Chemical Identifiers
| 1116-40-1 | |
| 185.36 | |
| IIFFFBSAXDNJHX-UHFFFAOYSA-N | |
| tris(2-methylpropyl)amine |
| C12H27N | |
| MFCD00040501 | |
| 14222 | |
| CC(C)CN(CC(C)C)CC(C)C |
Specifications
| 1116-40-1 | |
| 183°C | |
| MFCD00040501 | |
| 2734 | |
| CC(C)CN(CC(C)C)CC(C)C | |
| 185.36 | |
| 185.36 | |
| Liquid |
| Yellow | |
| C12H27N | |
| 25 mL | |
| IIFFFBSAXDNJHX-UHFFFAOYSA-N | |
| tris(2-methylpropyl)amine | |
| 14222 | |
| ≥98.0% (GC,T) | |
| Triisobutylamine |
Safety and Handling
TSCA : Yes